Anti-GALC Rabbit polyclonal antibody

Supplier: Biorbyt
ORB100385-100 ORB100385-20 ORB100385-200 ORB100385-5
BIRBORB100385-100EA 0 EUR
BIRBORB100385-100 BIRBORB100385-20 BIRBORB100385-200 BIRBORB100385-5
Anti-GALC Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: GALC (galactosylceramidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR