Anti-p95/NBS1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB105938-100 ORB105938-20
BIRBORB105938-100EA 0 EUR
BIRBORB105938-100 BIRBORB105938-20
Anti-p95/NBS1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000

Type: Primary
Antigen: p95/NBS1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR