Anti-FRS2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106513-100 ORB106513-20
BIRBORB106513-100EA 0 EUR
BIRBORB106513-100 BIRBORB106513-20
Anti-FRS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000

Type: Primary
Antigen: FRS2 (fibroblast growth factor receptor substrate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR