Anti-BRCA1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125493-100 ORB125493-200
BIRBORB125493-100EA 0 EUR
BIRBORB125493-100 BIRBORB125493-200
Anti-BRCA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR