Anti-DGKA Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125685-100 ORB125685-200
BIRBORB125685-100EA 0 EUR
BIRBORB125685-100 BIRBORB125685-200
Anti-DGKA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DGKA (diacylglycerol kinase, alpha 80kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR