Anti-HTR5A Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125870-100 ORB125870-200
BIRBORB125870-100EA 0 EUR
BIRBORB125870-100 BIRBORB125870-200
Anti-HTR5A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HTR5A (5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR