Anti-MT-ND1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126000-100 ORB126000-200
BIRBORB126000-100EA 0 EUR
BIRBORB126000-100 BIRBORB126000-200
Anti-MT-ND1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MT-ND1 (mitochondrially encoded NADH dehydrogenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR