Anti-MT-ND5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126001-100 ORB126001-200
BIRBORB126001-100EA 0 EUR
BIRBORB126001-100 BIRBORB126001-200
Anti-MT-ND5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MT-ND5 (mitochondrially encoded NADH dehydrogenase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR