Anti-BAX Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127747-100 ORB127747-200
BIRBORB127747-100EA 0 EUR
BIRBORB127747-100 BIRBORB127747-200
Anti-BAX Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR