Anti-HNRNPA1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128184-100 ORB128184-200
BIRBORB128184-100EA 0 EUR
BIRBORB128184-100 BIRBORB128184-200
Anti-HNRNPA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HNRNPA1 (heterogeneous nuclear ribonucleoprotein A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR