Anti-hnRNP C1/C2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128185-100 ORB128185-200
BIRBORB128185-100EA 0 EUR
BIRBORB128185-100 BIRBORB128185-200
Anti-hnRNP C1/C2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: hnRNP C1/C2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR