Anti-KCNA1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128246-100 ORB128246-200
BIRBORB128246-100EA 0 EUR
BIRBORB128246-100 BIRBORB128246-200
Anti-KCNA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCNA1 (potassium voltage-gated channel, shaker-related subfamily, member 1 (episodic ataxia with myokymia))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR