Anti-KCNA5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128247-100 ORB128247-200
BIRBORB128247-100EA 0 EUR
BIRBORB128247-100 BIRBORB128247-200
Anti-KCNA5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCNA5 (potassium voltage-gated channel, shaker-related subfamily, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR