Anti-TDG Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128823-100 ORB128823-200
BIRBORB128823-100EA 0 EUR
BIRBORB128823-100 BIRBORB128823-200
Anti-TDG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TDG (thymine-DNA glycosylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR