Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB129627-100 ORB129627-200
BIRBORB129627-100EA 0 EUR
BIRBORB129627-100 BIRBORB129627-200
Anti-HDAC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR