Anti-VEGF Receptor 3 Rabbit Polyclonal Antibody

Supplier: Abcam
AB27278-100 AB27278-500 AB27278-1
ABCAAB27278-100EA 588 EUR
ABCAAB27278-100 ABCAAB27278-500 ABCAAB27278-1
Anti-VEGF Receptor 3 Rabbit Polyclonal Antibody
Antibodies
Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: FLT4 (fms-related tyrosine kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR