Anti-Histone H2A (acetyl K5) Rabbit Monoclonal Antibody [clone: EP856Y]

Supplier: Abcam
AB45152-100 AB45152-1ML
ABCAAB45152-100EA 713 EUR
ABCAAB45152-100 ABCAAB45152-1ML
Anti-Histone H2A (acetyl K5) Rabbit Monoclonal Antibody [clone: EP856Y]
Antibodies
Rabbit monoclonal [EP856Y] to Histone H2A (acetyl K5).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H2A
Clonality: Monoclonal
Clone: EP856Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR