Anti-HSF1 Rabbit Monoclonal Antibody [clone: EP1710Y]

Supplier: Abcam
AB52757-100 AB52757-1ML
ABCAAB52757-100EA 688 EUR
ABCAAB52757-100 ABCAAB52757-1ML
Anti-HSF1 Rabbit Monoclonal Antibody [clone: EP1710Y]
Rabbit monoclonal [EP1710Y] to HSF1 - ChIP Grade.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: EP1710Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR