Anti-MUC1 Mouse Monoclonal Antibody [clone: HMFG1 (aka 1.10.F3)]

Supplier: Abcam
AB70475-100 AB70475-1MG
ABCAAB70475-100EA 713 EUR
ABCAAB70475-100 ABCAAB70475-1MG
Anti-MUC1 Mouse Monoclonal Antibody [clone: HMFG1 (aka 1.10.F3)]
Antibodies
Mouse monoclonal [HMFG1 (aka 1.10.F3)] to MUC1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: HMFG1 (aka 1.10.F3)
Conjugation:
Epitope: This antibody recognizes a peptide epitope (PDTR) within the VNTR region of the extracellular domain of MUC1 (PubMed ID: PMC3021526)
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR