Anti-Nicotinic Acetylcholine Receptor beta/CHRNB1 Rabbit Monoclonal Antibody [clone: EP2067Y]

Supplier: Abcam
AB76159-100 AB76159-1ML
ABCAAB76159-100EA 607 EUR
ABCAAB76159-100 ABCAAB76159-1ML
Anti-Nicotinic Acetylcholine Receptor beta/CHRNB1 Rabbit Monoclonal Antibody [clone: EP2067Y]
Antibodies
Rabbit monoclonal [EP2067Y] to Nicotinic Acetylcholine Receptor beta/CHRNB1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Nicotinic Acetylcholine Receptor beta/CHRNB1
Clonality: Monoclonal
Clone: EP2067Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR