Anti-Glucagon Rabbit Monoclonal Antibody [clone: EPR3070-45]

Supplier: Abcam
AB108426-100 AB108426-1ML
ABCAAB108426-100EA 475 EUR
ABCAAB108426-100 ABCAAB108426-1ML
Anti-Glucagon Rabbit Monoclonal Antibody [clone: EPR3070-45]
Antibodies
Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: GCG (glucagon)
Clonality: monoclonal
Clone: EPR3070
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR