Anti-Myosin light chain 3 Rabbit Monoclonal Antibody [clone: EPR4160]

Supplier: Abcam
AB108923-100 AB108923-1ML
ABCAAB108923-100EA 707 EUR
ABCAAB108923-100 ABCAAB108923-1ML
Anti-Myosin light chain 3 Rabbit Monoclonal Antibody [clone: EPR4160]
Antibodies
Rabbit monoclonal [EPR4160] to Myosin light chain 3.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Myosin light chain 3
Clonality: Monoclonal
Clone: EPR4160
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR