Anti-HDAC6 Rabbit Monoclonal Antibody [clone: EPR1698(2)]

Supplier: Abcam
AB133493-100 AB133493-20UL AB133493-1ML
ABCAAB133493-100EA 819 EUR
ABCAAB133493-100 ABCAAB133493-20UL ABCAAB133493-1ML
Anti-HDAC6 Rabbit Monoclonal Antibody [clone: EPR1698(2)]
Antibodies
Rabbit monoclonal [EPR1698(2)] to HDAC6.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HDAC6
Clonality: Monoclonal
Clone: EPR1698(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;African green monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR