Anti-Methionine Aminopeptidase 2/p67 Rabbit Monoclonal Antibody [clone: EPR6887]

Supplier: Abcam
AB134124-100 AB134124-1ML
ABCAAB134124-100EA 625 EUR
ABCAAB134124-100 ABCAAB134124-1ML
Anti-Methionine Aminopeptidase 2/p67 Rabbit Monoclonal Antibody [clone: EPR6887]
Antibodies
Rabbit monoclonal [EPR6887] to Methionine Aminopeptidase 2/p67.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Methionine Aminopeptidase 2/p67
Clonality: Monoclonal
Clone: EPR6887
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR