Anti-ANKRD28 Rabbit Monoclonal Antibody [clone: EPR9211]

Supplier: Abcam
AB137091-100 AB137091-1ML
ABCAAB137091-100EA 700 EUR
ABCAAB137091-100 ABCAAB137091-1ML
Anti-ANKRD28 Rabbit Monoclonal Antibody [clone: EPR9211]
Antibodies
Rabbit monoclonal [EPR9211] to ANKRD28.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ANKRD28
Clonality: Monoclonal
Clone: EPR9211
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR