Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20112 20112-1
20112.EA 300 EUR
20112. 20112-1
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR