Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium
20120 20120-1
20120.EA 300 EUR
20120. 20120-1
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR