Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20159 20159-1
20159.EA 359 EUR
20159. 20159-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR