Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20164 20164-1
20164.EA 300 EUR
20164. 20164-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR