Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)

Supplier: Biotium
20172 20172-1
20172.EA 359 EUR
20172. 20172-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR