Anti-IgG Chicken Polyclonal Antibody (CF™ 488A)

Supplier: Biotium
20225-1 20225
20225-1EA 127 EUR
20225-1 20225.
Anti-IgG Chicken Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR