Anti-IgG Chicken Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20226 20226-1
20226.EA 300 EUR
20226. 20226-1
Anti-IgG Chicken Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Goat
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 594
Reactivity Goat
Host Chicken
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR