Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium
20236 20236-1
20236.EA 300 EUR
20236. 20236-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR