Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium
20329 20329-1
20329.EA 249 EUR
20329. BTIU20329-1
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™543 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR