Anti-NBS1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-143 NB100-143SS
NOVUNB100-143EA 695 EUR
NOVUNB100-143 NOVUNB100-143SS
Anti-NBS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NBS1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR