Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]

Supplier: Novus Biologicals
NB100-168 NB100-168SS
NOVUNB100-168EA 596 EUR
NOVUNB100-168 NOVUNB100-168SS
Anti-MGMT Mouse Monoclonal Antibody [clone: MT 23.2]
Antibodies
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: monoclonal
Clone: MT 23.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR