Anti-Mitofilin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1919 NB100-1919SS
NOVUNB100-1919EA 655 EUR
NOVUNB100-1919 NOVUNB100-1919SS
Anti-Mitofilin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Mitofilin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR