Anti-SYVN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2526 NB100-2526SS
NOVUNB100-2526EA 526 EUR
NOVUNB100-2526 NOVUNB100-2526SS
Anti-SYVN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SYVN1 (synovial apoptosis inhibitor 1, synoviolin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR