Anti-LOX propeptide Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-41568 NB110-41568SS
NOVUNB110-41568EA 606 EUR
NOVUNB110-41568 NOVUNB110-41568SS
Anti-LOX propeptide Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LOX propeptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR