Anti-TMED10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNB110-57587EA 542 EUR
NOVUNB110-57587 NOVUNB110-57587SS
Anti-TMED10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR