Anti-APBB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-58360 NB110-58360SS
NOVUNB110-58360EA 554 EUR
NOVUNB110-58360 NOVUNB110-58360SS
Anti-APBB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR