Anti-TNC Mouse Monoclonal Antibody [clone: 4C8MS]

Supplier: Novus Biologicals
NB110-68136 NB110-68136SS
NOVUNB110-68136EA 702 EUR
NOVUNB110-68136 NOVUNB110-68136SS
Anti-TNC Mouse Monoclonal Antibody [clone: 4C8MS]
Antibodies
Type: Primary
Antigen: TNC (tenascin C)
Clonality: monoclonal
Clone: 4C8MS
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR