Anti-TRPM2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-81601 NB110-81601SS
NOVUNB110-81601EA 642 EUR
NOVUNB110-81601 NOVUNB110-81601SS
Anti-TRPM2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRPM2 (transient receptor potential cation channel, subfamily M, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR