Anti-SPP1 Mouse Monoclonal Antibody [clone: 1B20]

Supplier: Novus Biologicals
NB110-89062 NB110-89062SS
NOVUNB110-89062EA 640 EUR
NOVUNB110-89062 NOVUNB110-89062SS
Anti-SPP1 Mouse Monoclonal Antibody [clone: 1B20]
Antibodies
The Osteopontin / OPN Antibody (1B20) from Novus Biologicals is a mouse monoclonal antibody to Osteopontin / OPN. This antibody reacts with human, rat, rabbit. The Osteopontin / OPN Antibody (1B20) has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: OPN
Clonality: Monoclonal
Clone: 1B20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR