Anti-HIST3H3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB21-1142 NB21-1142SS
NOVUNB21-1142EA 612 EUR
NOVUNB21-1142 NOVUNB21-1142SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR