Anti-HIST4H4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB21-2044 NB21-2044SS
NOVUNB21-2044EA 526 EUR
NOVUNB21-2044 NOVUNB21-2044SS
Anti-HIST4H4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR