Anti-Sodium Potassium ATPase Beta 1 Mouse Monoclonal Antibody [clone: 464.8]

Supplier: Novus Biologicals
NB300-147 NB300-147SS
NOVUNB300-147EA 570 EUR
NOVUNB300-147 NOVUNB300-147SS
Anti-Sodium Potassium ATPase Beta 1 Mouse Monoclonal Antibody [clone: 464.8]
Antibodies
Type: Primary
Antigen: Sodium Potassium ATPase Beta 1
Clonality: monoclonal
Clone: 464.8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol ATP1B1
Antigen name Sodium Potassium ATPase Beta 1
Environmentally Preferable
Antigen synonyms ATP1B
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 464.8
Reactivity Human
Host Mouse
Gene ID 481
Isotype IgG2a kappa
Western blot Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR