Anti-CTDSPL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-232 NB300-232SS
NOVUNB300-232EA 732 EUR
NOVUNB300-232 NOVUNB300-232SS
Anti-CTDSPL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTDSPL (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol SCP3
Antigen name Synaptonemal Complex Protein 3
Environmentally Preferable
Antigen synonyms Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 50511
Isotype
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR