Anti-LXR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB400-157 NB400-157SS
NOVUNB400-157EA 626 EUR
NOVUNB400-157 NOVUNB400-157SS
Anti-LXR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LXR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR