Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-236 NB500-236SS
NOVUNB500-236EA 606 EUR
NOVUNB500-236 NOVUNB500-236SS
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR